Learn More
Medchemexpress LLC HY-12342 10mg , ML216 CAS:1430213-30-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1234210MG
Encompass_Preferred
HY-12342 10mg ML216 CAS: 1430213-30-1 ML216 (CID-49852229) is a potent, selective and cell permeable inhibitor of the DNA unwinding activity of BLM helicase with IC50s of 2.98 μM and 0.97 μM for BLMfull-length and BLM636-1298, respectively. ML216 inhibits ssDNA-dependent ATPase activity of BLM with a Ki of 1.76 µM. Antitumor avtivity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0620