Learn More
TARGETMOL CHEMICALS INC ML228 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T783610MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ML228 is a potent the Hypoxia Inducible Factor (HIF) pathway activator with EC50 of 1 uM. ML228 potently activates HIF in vitro as well as its downstream target VEGF. purity: 99%
Catalog No. 50-257-9117