Learn More
Medchemexpress LLC ML228 | 1357171-62-0 | 98.0% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1275410MG
Encompass_Preferred
ML228 is a potent Hypoxia Inducible Factor (HIF) pathway activator with an EC50 of 1 μM. It potently activates HIF in vitro as well as its downstream target VEGF. ML228 represents a novel chemotype for the study of HIF activation and its therapeutic potential.
  • Potently activates HIF in vitro
  • Activates downstream target VEGF
  • Novel chemotype for HIF activation studies
  • Structurally different from known HIF activators
  • Lacks acidic functional group found in PHD inhibitors
  • Improves local hypoxic ischemia environment in vivo
  • Reduces SCI secondary injury in vivo
  • Promotes recovery of neurological function in SD rats
Catalog No. 50-003-42445