Learn More
Medchemexpress LLC ML230 | 1776055-05-0 | 99.65% | 439.47 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY111678100MG
Encompass_Preferred
ML230 is a selective inhibitor of ATP-binding cassette (ABC) transporter ABCG2.
  • Demonstrates 36-fold selectivity for ABCG2 over ABCB1.
  • EC50 values of 0.13 μM for ABCG2 and 4.65 μM for ABCB1.
Catalog No. 50-004-40083