Learn More
Medchemexpress LLC Ml230 | 1776055-05-0 | 99.7% | 439.47 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11167810MM/1ML
Encompass_Preferred
ML230 is a selective inhibitor of ATP-binding cassette (ABC) transporter ABCG2. It is 36-fold selective for ABCG2 over ABCB1 with EC50 values of 0.13 μM and 4.65 μM, respectively. It is intended for research use only.
  • Selective inhibitor of ATP-binding cassette transporter ABCG2.
  • 36-fold selective for ABCG2 over ABCB1.
  • Available in solid and solution forms.
  • Multiple quantities available.
  • Detailed solubility and storage information provided.
  • Information for *in vitro* and *in vivo* use.
Catalog No. 50-004-38959