Learn More
TARGETMOL CHEMICALS INC ML240 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T353510MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ML240 is a selective ATP-competitive p97 inhibitor. purity: 99%
Catalog No. 50-257-6921