Learn More
TARGETMOL CHEMICALS INC ML264 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T689410MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. ML264 (CID-51003603) is a selective kruppel-like factor 5 (KLF5) inhibitor, which potently Inhibits growth of Colorectal Cancer. Purity 98.72%
Catalog No. 50-287-7323