Learn More
Selleck Chemical LLC ML277 S0050-25MG
SDP
Supplier:  Selleck Chemical LLC S005025MG
Encompass_Preferred
ML277 (CID-53347902) is a potent activator of KCNQ1 channels with an EC50 of 260 nM and shows 100-fold selectivity over KCNQ2 and KCNQ4
Catalog No. 50-000-39772