Learn More
Medchemexpress LLC ML303 | 1638211-04-7 | 99.6% | 399.37 g·mol⁻1 | C21H16F3N3O2 | 25MG

Supplier: Medchemexpress LLC HY12613625MG
ML303 is a pyrazolopyridine small-molecule antagonist of influenza virus nonstructural protein 1 (NS1), reported with an IC90 of 155 nM and an antiviral EC50 of 0.7 μM against Influenza A (H1N1). It is provided as a high-purity research compound intended for biochemical and cellular assays investigating NS1 function and viral replication.
- Pyrazolopyridine NS1 antagonist with reported IC90 155 nM and EC50 0.7 μM vs Influenza A (H1N1).
- Molecular formula C21H16F3N3O2 and molecular weight 399.37 g·mol⁻1.
- Purity 99.6%.
- Available as a powder in multiple small pack sizes for research use.
- Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
- Intended for research use only; follow the safety data sheet for handling.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.