Learn More
Medchemexpress LLC Ml311 | 315698-17-0 | 99.0% | 415.45 | C23H24F3N3O | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1017781MG
Encompass_Preferred
ML311 is a small-molecule research compound that selectively inhibits the Mcl-1/Bim protein-protein interaction and is used in cellular and in vivo pharmacology studies. Reported data show submicromolar cellular potency across multiple cancer cell lines. The product is supplied as a high-purity solid with validated solubility and storage recommendations for preclinical use.
  • Potent and selective inhibitor of the Mcl-1/Bim interaction.
  • High purity (approximately 99%).
  • White to off-white solid suitable for analytical and biological assays.
  • DMSO soluble with published in vitro and in vivo formulation protocols.
  • Reported submicromolar EC50/GI50 activity across multiple cell lines.
  • Packaged in multiple milligram quantities for preclinical studies.
Catalog No. 50-002-49066