Learn More
Selleck Chemical LLC ML346
SDP
Supplier:  Selleck Chemical LLC S65682MG
Encompass_Preferred
ML346 is a Hsp70 activator and a novel modulator of proteostasis for protein conformational diseases
Catalog No. 50-000-02925