Learn More
Apexbio Technology LLC ML347, 5mg. Cas: 1062368-49-3 MFCD: MFCD28099805. BMP receptor inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC B36885
Encompass_Preferred
ML347 is a potent and selective inhibitor of bone morphogenetic protein (BMP) receptor with IC50 value of 32nM against ALK2 [1].ML347 is discovered as a selective inhibitor of BMP type-I receptor ALK2 versus ALK3 and is identified as a probe molecule. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5350