Learn More
Selleck Chemical LLC ML348-5mg
SDP
Supplier:  Selleck Chemical LLC S65645MG
Encompass_Preferred
ML348 (GNF-Pf-1127) is a potent and selective APT1 (Acyl protein thioesterase 1)/lysophospholipase 1 (LYPLA1) inhibitor with Ki values of 280 nM and >10 μM for APT1 and APT2, respectively.
Catalog No. 50-238-9647