Learn More
Selleck Chemical LLC ML351 S3387-5mg
SDP
Supplier:  Selleck Chemical LLC S33875MG
Encompass_Preferred
ML351 is a selective inhibitor of 15-Lipoxygenase-1 (15-LOX-1 or 12/15-LOX) with IC50 of 200 nM
Catalog No. 50-310-9313