Learn More
Medchemexpress LLC ML351 | 847163-28-4 | 99.2% | 249.27 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11131010MG
Encompass_Preferred
ML351 is a potent and highly specific 15-LOX-1 inhibitor with an IC50 of 200 nM. It demonstrates excellent selectivity (>250-fold) over related isozymes such as 5-LOX, platelet 12-LOX, 15-LOX-2, ovine COX-1, and human COX-2. ML351 has been shown to prevent dysglycemia and reduce β-cell oxidative stress in a nonobese diabetic mouse model of T1D.
  • Potent and highly specific 15-LOX-1 inhibitor with an IC50 of 200 nM.
  • Exhibits excellent selectivity (>250-fold) against related isozymes, including 5-LOX, platelet 12-LOX, 15-LOX-2, ovine COX-1, and human COX-2.
  • Prevents dysglycemia and reduces β-cell oxidative stress in a nonobese diabetic mouse model of T1D.
  • Protects against diabetes development in an STZ β-cell injury model.
  • Leads to improved glycemic control and significantly reduced insulitis in NOD mice.
Catalog No. 50-003-43303