Learn More
Selleck Chemical LLC ML355
SDP
Supplier:  Selleck Chemical LLC S65575MG
Encompass_Preferred
ML355 is a potent and selective inhibitor of human 12-Lipoxygenase with an IC50 of 290 nM and shows excellent selectivity over related lipoxygenases and cyclooxygenases
Catalog No. 50-000-02825