Learn More
Selleck Chemical LLC ML367 S0161-25MG
SDP
Supplier:  Selleck Chemical LLC S016125MG
Encompass_Preferred
ML367 is a potent micromolar inhibitor of ATPase family AAA domain-containing protein 5 (ATAD5) stabilization ML367 blocks DNA repair pathways and general DNA damage responses including RPA32-phosphorylation and CHK1-phosphorylation in response to UV irradiation
Catalog No. 50-000-39361