Learn More
Medchemexpress LLC HY-122198 50mg Medchemexpress, ML367 CAS:381168-77-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12219850MG
Encompass_Preferred
Medchemexpress, HY-122198 50mg ML367 CAS:381168-77-0 ML367 is a potent inhibitor of ATPase family AAA domain-containing protein 5 (ATAD5) stabilization, acts as a probe molecule that has low micromolar inhibitory activity. ML367 blocks DNA repair pathways, suppresses general DNA damage responses including RPA32-phosphorylation and CHK1-phosphorylation in response to UV irradiation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0983