Learn More
Medchemexpress LLC HY-12439 5mg Medchemexpress, ML380 CAS:1627138-52-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY124395MG
Encompass_Preferred
Medchemexpress, HY-12439 5mg ML380 CAS:1627138-52-6 ML380 is a potent, subtype-selective, and brain-penetrant positive allosteric modulator (PAM) of M5 mAChR, with EC50s of 190 and 610 nM for human and rat M5, respectively. ML380 exhibits moderate selectivity versus the M1 and M3 mAChR subtypes. ML380 could increase the affinity of ACh for the M5 mAChR. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1044