Learn More
Medchemexpress LLC Ml382 | 1646499-97-9 | 99.89% | 360.43 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY11028510MG
Encompass_Preferred
ML382 is a potent and selective positive allosteric modulator of MRGPRX1 (Mas-related G protein-coupled receptor X1, MrgX1), with an EC50 of 190 nM. It is intended for research use only and not sold to patients. In vitro, ML382 dose-dependently increases the potency of BAM8-22, a MRGPRX1 agonist. In vivo, ML382 increases inhibition of ICa by BAM8-22 in DRG neurons from MrgprX1 mice, enhances the inhibition of spinal synaptic transmission by BAM8-22, attenuates heat hypersensitivity, and inhibits nerve injury-induced ongoing pain in MrgprX1 mice.
  • Potent and selective positive allosteric modulator of MRGPRX1
  • Increases potency of BAM8-22 in vitro
  • Enhances inhibition of spinal synaptic transmission in vivo
  • Attenuates heat hypersensitivity
  • Inhibits nerve injury-induced ongoing pain
Catalog No. 50-003-43097