Learn More
Cayman Chemical ML9 Inhibitors
SDP
Supplier:  Cayman Chemical 10010236100 MG
Encompass_Preferred
An inhibitor of PKB/Akt activity (IC50 = 10-50 µM) that inhibits insulin-stimulated glucose transport and intracellular protein translocation; inhibits additional serine/threonine kinases including PKA (IC50 = ~20 µM), p90 S6 (IC50 = ~50 µM), and MAP kinase (IC50 = ~35 µM)
Catalog No. 50-289-1168