Learn More
Sino Biological Peptide Substrates: MLC Peptide
Supplier:  Sino Biological M0958
Encompass_Preferred
The MLC Peptide sequence (AKRPQRATSNVFS) represents the phosphate-accepting domain of human MYL9 (amino acids 12-23) and can be used as a substrate for PHKG1, PHKG2 and DAPK2.
Catalog No. 50-256-8693