Learn More
Santa Cruz Biotechnology MLGGBPHRP
Supplier:  Santa Cruz Biotechnology SC516102
Encompass
Mouse IgG kappa binding protein (m-IgGκ BP) conjugated to Horseradish Peroxidase (HRP) * supplied at 200 µg in 0.5 ml volume * Strongly recommended alternative to conventional anti-mouse IgG secondary antibodies for Western blotting (WB) and Immunohistochemistry (IHC) * Suitable for binding to mouse IgGκ light chain immunoglobulins, comprising approximately 98% of mouse monoclonal antibodies; not suitable for use with mouse monoclonal IgGλ light chain antibodies * D177Highly specific reagent that provides strong signal with minimal background and virtually complete elimination of lot to lot variation associated with conventionally generated secondary antibodies
Catalog No. NC1187299