Learn More
ABclonal Technology MLH3 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2550420UL
Encompass_Preferred
This gene is a member of the MutL-homolog (MLH) family of DNA mismatch repair (MMR) genes MLH genes are implicated in maintaining genomic integrity during DNA replication and after meiotic recombination The protein encoded by this gene functions as a heterodimer with other family members Somatic mutations in this gene frequently occur in tumors exhibiting microsatellite instability and germline mutations have been linked to hereditary nonpolyposis colorectal cancer type 7 (HNPCC7) Several alternatively spliced transcript variants have been identified but the full-length nature of only two transcript variants has been determined
Catalog No. 50-000-11022