Learn More
Selleck Chemical LLC MLi-2 5mg
SDP
Supplier:  Selleck Chemical LLC S96945MG
Encompass_Preferred
MLi-2 is an orally active and highly selective inhibitor ofLRRK2. MLi-2 exhibits exceptional potency in a purified LRRK2 kinase assay in vitro with IC50 of 0.76 nM, a cellular assay monitoring dephosphorylation of LRRK2 pSer935 LRRK2 with IC50 of 1.4 nM, and a radioligand competition binding assay with IC50 of 3.4 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3496