Learn More
TARGETMOL CHEMICALS INC MLN-4760 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T161165MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. MLN-4760 is an angiotensin-converting (ACE2) inhibitor(human ACE2;IC50=0.44 nM). It also has excellent selectivity (>5000-fold) versus related enzymes including human testicular ACE (IC50 >100 uM) and bovine carboxypeptidase A (CPDA; IC50 27 uM). purity: 99%
Catalog No. 50-258-1190