Learn More
Cayman Chemical MLN4924, 905579-51-3, 1 mg
SDP
Supplier:  Cayman Chemical 152171
Encompass_Preferred
Molecular Formula C21H25N5O4S, Purity ≥98%, Formula Weight 443.5, Synonyms: Pevonedistat; An analog of adenosine 5'-monophosphate that selectively inhibits NAE (IC50 = 4.7 nM) and disrupts CRL-mediated protein turnover leading to apoptosis in cancer cells by deregulating S-phase DNA synthesis.
Catalog No. NC0630547