Learn More
AdipoGen MLN4924 [NAE Inhibitor] Solution,Chemical. CAS: 905579-51-3
SDP
Supplier:  AdipoGen AGCS10103M001
Encompass_Preferred
MLN4924 [NAE Inhibitor] Solution, AdipoGen Life Sciences. Chemical. CAS: 905579-51-3. Formula: C21H25N5O4S. MW: 443.5. Potent, selective and cell-permeable inhibitor of Nedd8 E1 Activating Enzyme (NAE). By inhibiting the Neddylation pathway this molecule disrupts cullin-RING ligase activity, leading to inhibition of proliferation or apoptotic death of various human tumor cells through the deregulation of S-phase DNA synthesis.
Catalog No. 50-114-6629