Learn More
Selleck Chemical LLC MLN8054 50mg 869363-13-3
SDP
Supplier:  Selleck Chemical LLC S110050MG
Encompass_Preferred
MLN8054 is a potent and selective inhibitor of Aurora A with IC50 of 4 nM in Sf9 insect cell. It is more than 40-fold selective for Aurora A than Aurora B. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-853-1