Learn More
Apexbio Technology LLC MLR 1023 41964-07-2 5mg
SDP
Supplier:  Apexbio Technology LLC B56485
Encompass_Preferred
MLR 1023 (CAS 41964-07-2) also known as Tolimidone functions as an allosteric activator of Lyn kinase by binding to a non-catalytic site on the target protein thereby enhancing kinase activity In vitro studies indicate an EC50 value of approximately 63 nM for the allosteric activation of Lyn kinase MLR 1023 is widely utilized as a research tool to elucidate Lyn kinase-mediated signaling cellular functions and related pathological mechanisms Its application extends to investigations of kinase pathway modulation and potential pharmacological intervention points in disease models
Catalog No. 50-000-88577