Learn More
Medchemexpress LLC MLT-747 | 2097853-86-4 | 99.6% | 478.33 | C20H21Cl2N7O3 | 50MG
SDP
Supplier:  Medchemexpress LLC HY12458750MG
Encompass_Preferred
MLT-747 is a potent, selective, allosteric inhibitor of MALT1 that binds the Trp580 allosteric pocket and inhibits MALT1 protease activity with an IC50 of 14 nM. It is used as a research tool to probe MALT1-dependent signaling and downstream NF-κB activation in biochemical and cellular studies.
Catalog No. 50-002-39355