Learn More
Medchemexpress LLC MLT-748 100mg | 1832578-30-9 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY115466100MG
Encompass_Preferred
MLT-748 is a potent, selective, allosteric inhibitor of MALT1 that binds the Trp580 pocket and inhibits protease activity with an IC50 of ~5 nM. It is supplied for biochemical and cellular studies to probe MALT1-dependent signaling and preclinical research.
  • Potent MALT1 inhibition (IC50 ~5 nM).
  • Allosteric mechanism by binding the Trp580 pocket.
  • High purity suitable for research applications.
  • Available as solid or DMSO solution for assay convenience.
  • Stable under recommended storage conditions for long-term use.
Catalog No. 50-003-07914