Learn More
Medchemexpress LLC MLT-748 50mg | 1832578-30-9 | 492.32 | C19H19Cl2N9O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11546650MG
Encompass_Preferred
MLT-748 is a research-grade small-molecule allosteric inhibitor of MALT1 for use in biochemical and cellular research. It binds the Trp580 allosteric pocket and shows an IC50 of 5 nM. Supplied in high-purity solid and solution formats for laboratory assays.
  • Potent, selective allosteric inhibition of MALT1 (IC50 = 5 nM).
  • Binds the Trp580 allosteric pocket on MALT1.
  • High purity suitable for research (reported purity ≈ 99.89%).
  • Available as a solid and as a 10 mM solution in DMSO for convenience.
  • Suitable for biochemical assays and cellular studies of MALT1 function.
  • Molecular weight 492.32 and formula C19H19Cl2N9O3; CAS 1832578-30-9.
Catalog No. 50-002-29495