Learn More
Medchemexpress LLC HY-108003 10mg Medchemexpress, MM 07 CAS:1876450-21-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10800310MG
Encompass_Preferred
Medchemexpress, HY-108003 10mg MM 07 CAS:1876450-21-3 MM 07 is a biased apelin receptor agonist, with a KD of 300 nM in CHO-K1 cells and a KD of 172 nM in human heart. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8497