Learn More
Abcam MM 11253, RARgamma, 50MG
Supplier:  Abcam AB14150850MG
Encompass_Preferred
MW 462.66 g/mol, Purity >98%. Selective RARγ antagonist. Blocks the growth inhibitory effects of RARγ-selective agonists. Shows antiapoptotic effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6548