Learn More
Selleck Chemical LLC MMAE
SDP
Supplier:  Selleck Chemical LLC S7721100MG
Encompass_Preferred
MMAE (Monomethyl auristatin E) is a synthetic antineoplastic agent It is also a microtubule-disrupting agent
Catalog No. 50-000-03798