Learn More
Medchemexpress LLC HY-15579 100mg , MMAF CAS:745017-94-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15579100MG
Encompass_Preferred
HY-15579 100mg MMAF CAS: 745017-94-1 MMAF (Monomethylauristatin F) is a potent tubulin polymerization inhibitor and is used as a antitumor agent. MMAF (Monomethylauristatin F) is widely used as a cytotoxic component of antibody-drug conjugates (ADCs) such as vorsetuzumab mafodotin and SGN-CD19A. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0922