Learn More
Selleck Chemical LLC MMAF S0328-10MM/1ML
SDP
Supplier:  Selleck Chemical LLC S032810MM/1ML
Encompass_Preferred
MMAF (Monomethylauristatin F) is a synthetic antineoplastic agent and a potent inhibitor of tubulin polymerization MMAF (Monomethylauristatin F) is applied as a cytotoxic component of antibody-drug conjugates (ADCs) such as vorsetuzumab mafodotin ABT-414 and SGN-CD19A
Catalog No. 50-000-43567