Learn More
Medchemexpress LLC MMAF intermediate 2 | 864238-20-0 | 99.8% | C19H29ClN2O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY79254
Encompass_Preferred
MMAF intermediate 2 is an intermediate used in the synthesis of MMAF (Monomethylauristatin F). MMAF is a potent inhibitor of tubulin polymerization and serves as the cytotoxic component in classic antibody-drug conjugates (ADCs), such as Vorsetuzumab mafodotin and SGN-CD19A.
  • Potent inhibitor of tubulin polymerization
  • Cytotoxic component in antibody-drug conjugates (ADCs)
  • Used in the synthesis of Monomethylauristatin F (MMAF)
  • Appears as a white to off-white solid
Catalog No. 50-003-57534