Learn More
Medchemexpress LLC MMAF-OtBu | 745017-95-2 | 99.8% | 788.07 | C43H73N5O8 | 1 ML

Supplier: Medchemexpress LLC HY15622510MM 1ML
MMAF-OtBu is a modified monomethylauristatin F derivative used as a cytotoxic payload in antibody-drug conjugate (ADC) synthesis and related research applications. It is supplied as a solid or as a ready-to-use 10 mM solution in DMSO (1 mL), with high reported purity and characterized identity.
- Supplied as a 10 mM solution in DMSO (1 mL) and as solids in multiple sizes.
- Available solid sizes: 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, with larger sizes by quote.
- High purity (reported 99.83%).
- Molecular weight 788.07; chemical formula C43H73N5O8; CAS 745017-95-2.
- Intended for research use only; not for human therapeutic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.