Learn More
Medchemexpress LLC MMG-11 | 313254-94-3 | 306.27 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY112146100MG
Encompass_Preferred
MMG-11 is a potent and selective human TLR2 antagonist with low cytotoxicity. It inhibits both TLR2/1 and TLR2/6 signaling with IC50s of 1.7 μM for Pam3CSK4-induced hTLR2/1 and 5.7 μM for Pam2CSK4-induced hTLR2/6 responses.
  • Potent and selective human TLR2 antagonist.
  • Low cytotoxicity.
  • Inhibits both TLR2/1 and TLR2/6 signaling with IC50s of 1.7 μM for Pam3CSK4-induced hTLR2/1 and 5.7 μM for Pam2CSK4-induced hTLR2/6 responses.
  • Does not show cellular toxicity or interference with signaling induced by other TLR agonists, IL-1β or TNF.
  • Exhibits no cytotoxic effects up to 100 μM in peripheral blood mononuclear cells (PBMCs).
  • Effective inhibition of TLR2/1 and TLR2/6 signaling.
Catalog No. 50-004-37080