Learn More
Medchemexpress LLC HY-112146A 10mg Medchemexpress, MMG-11 (quarterhydrate) CAS:0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY112146A10MG
Encompass_Preferred
Medchemexpress, HY-112146A 10mg MMG-11 (quarterhydrate) CAS:0 MMG-11 quarterhydrate is a potent and selective human TLR2 antagonist with low cytotoxicity. MMG-11 quarterhydrate inhibits both TLR2/1 and TLR2/6 signaling with IC50s of 1.7 µM for Pam3CSK4-induced hTLR2/1 and 5.7 µM for Pam2CSK4-induced hTLR2/6 responses. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9716