Learn More
Apexbio Technology LLC MMK 1 1mg
SDP
Supplier:  Apexbio Technology LLC B54461
Encompass_Preferred
MMK 1 (CAS 271246-66-3) is a highly selective agonist of formyl peptide receptor-like 1 (FPRL-1) exhibiting subnanomolar half-maximal effective concentration (EC50 2 nM) for FPRL-1 while showing minimal activity against FPR1 (EC50 10 000 nM) By engaging FPRL-1 MMK 1 induces chemotaxis and calcium mobilization in phagocytic leukocytes and enhances interleukin-1 (IL-1 ) production in human monocytes sensitive to pertussis toxin Its ability to modulate innate immune cell functions makes MMK 1 a valuable tool for investigating mechanisms of inflammation immune regulation and associated behavioral responses such as anxiety in preclinical research
Catalog No. 50-000-90389