Learn More
Enzo Life Sciences MMP-11 (catalytic domain) (human), (recombinant) (10 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE2820010
Encompass_Preferred
Alternative name: Matrix metalloproteinase 11. Stromelysin 3. MW: 19.3 kDa. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 50mM TRIS, 5mM CaCl2, 300mM NaCl, 20µM ZnCl2, 0.5% Brij-35, and 30% glycerol. Source: Produced in E. coli. Active Matrix Metalloproteinase-11 (MMP-11, Stromelysin-3) catalytic domain from human cDNA. The enzyme consists of the catalytic domain of human MMP-11 (Phe98-Ser266, NM_005940) with a C-terminal purification tag. MMPs lacking this domain cannot cleave native collagens; however, activity toward other targets such as gelatin. CASein, or peptide substrates is unaffected. It may be an important link between obesity and cancer. UniProt: P24347. Long Term Storage: -80°C
Catalog No. NC1630288