Learn More
Enzo Life Sciences MMP-13 (catalytic domain) (human), (recombinant) (10 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE2460010
Encompass_Preferred
Alternative name: Matrix metalloproteinase 13. Collagenase 3. MW: 20.4 kDa. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 50mM TRIS, 5mM CaCl2, 300mM NaCl, 20µM ZnCl2, 0.5% Brij-35, and 30% glycerol. Source: Produced in E. coli. Active Matrix Metalloproteinase-13 (MMP-13, collagenase-3) catalytic domain from human cDNA. The enzyme consists of the catalytic domain of human MMP-13 (Tyr104-Asn274, NM_002427) with a C-terminal purification tag. This represents a naturally-occurring active form of MMP-13 which lacks the C-terminal hemopexin domain. MMPs lacking this domain cannot cleave native collagens; however, activity toward other targets such as gelatin. CASein, or peptide substrates is unaffected. UniProt: P45452. Long Term Storage: -80°C
Catalog No. NC1630290