Learn More
CPC Scientific MCA-Pro-Leu-Ala-Cys(p-OMeBz)-Trp-Ala-Arg(Dpa)-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific MMPS017B
Encompass
First synthetic substrate identified for cleavage by ST3. This fluorogenic peptide provides a simple test for routine assay of ST3 and is useful for ST3 inhibitor screens.ONE-LETTER SEQUENCE: Mca-PLAC(p-OMeBz)-WAR(Dpa)-NH2MOLECULAR FORMULA: C66H82N16O17S1MOLECULAR WEIGHT:1403.5STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1926163-82-7], SYNONYMS: MT1-MMP Substrate I, Fluorogenic, Matrix Metalloproteinase-14 Fluorogenic Substrate I, Mca-Pro-Leu-Ala-Cys(p-OMeBz)-Trp-Ala-Arg(Dpa)-NH2, Mca-PLAC(p-OMeBz)-WAR(Dpa)-NH2RESEARCH AREA: CancerREFERENCES: A. Mucha, et al., J. Biol. Chem. 273, 2763 (1998)
Catalog No. 50-210-9782