Learn More
Medchemexpress LLC MMP-2 Rat P. pasto 100ug
SDP
Supplier:  Medchemexpress LLC HYP700573100UG
Encompass_Preferred
MMP-2 is a multifunctional metalloproteinase that plays multiple roles in vasculature remodeling angiogenesis tissue repair tumor invasion inflammation and atherosclerotic plaque rupture In addition to degrading extracellular matrix proteins MMP-2 also acts on non-matrix proteins to promote vasoconstriction MMP-2 Protein Rat (P pastoris His) is the recombinant rat-derived MMP-2 protein expressed by P pastoris with N-6 His labeled tag
Catalog No. 50-001-74433