Learn More
Enzo Life Sciences MMP-3 fluorogenic substrate (0.1mg)
SDP
Supplier:  Enzo Life Sciences BMLP2780100
Encompass_Preferred
Highly-quenched, ultra-bright fluorogenic substrate for MMP-3 (other MMPs not tested, but likely).  5’-FAM fluorescence is thoroughly quenched by the TQ2W group until cleavage by MMP-3 separates the two moieties. Purity: ≥90% (HPLC). Solubility: Soluble in DMSO (5mM). Long Term Storage: -20°C.
Catalog No. 50-201-1331