Learn More
CPC Scientific DNP-Pro-Tyr-Ala-Tyr-Trp-Met-Arg-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific MMPS012B
Encompass
ONE-LETTER SEQUENCE: DNP-PYAYWMRMOLECULAR FORMULA: C54H65N13O14S1MOLECULAR WEIGHT:1152.25STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [158841-76-0], RESEARCH AREA: CancerREFERENCES: S. Netzel-Arnett, et al., Anal. Biochem. 195, 86 (1991)
Catalog No. 50-210-9772