Learn More
Selleck Chemical LLC MMP-9-IN-1 S0769-10MM/1ML
SDP
Supplier:  Selleck Chemical LLC S076910MM/1ML
Encompass_Preferred
MMP-9-IN-1 (OUN87710) is a specific inhibitor of matrix metalloproteinase-9 (MMP-9) MMP-9-IN-1 selectively targets the hemopexin (PEX) domain of MMP-9 with Kd of 2 1 M
Catalog No. 50-000-42572